1-Hexanesulphonic acid sodium salt, LiChropur™ for ion pair chromatography, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

1.18305.0025 1.18305.0100
1.18305.0025EA 468.79 EUR
1.18305.0025 1.18305.0100
1-Hexanesulphonic acid sodium salt, LiChropur™ for ion pair chromatography, Supelco®
1-Hexanesulphonic acid sodium salt
Formule: C₆H₁₃O₃SNa
Smeltpunt: 300 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007542
CAS nummer: 2832-45-3
EINECS: 220-601-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aebc3cb7a7cefac9ad5a7678d82a90d244b508f9&vid=d608216fe1e698d6b206b03588e6d6fe5ee661e1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Hill Formula C₆H₁₃NaO₃S
Molar Mass 188.22 g/mol
pH value 5.5 - 7.5 (100 g/l, H₂O, 20 °C)
Storage class 10 - 13 Other liquids and solids
WGK WGK 1 slightly water endangering


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR