1,4-Dithioerythritol, Millipore®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing


M-Clarity™ quality level = MQ200

1.24511.0005 1.24511.0025
1.24511.0005EA 102 EUR
1.24511.0005 1.24511.0025
1,4-Dithioerythritol, Millipore®
1,4-Dithioerythritol
Formule: C₄H₁₀O₂S₂
Kookpunt: 286 °C (1013 hPa)
Smeltpunt: 82…83 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00063750
CAS nummer: 6892-68-8
EINECS: 229-998-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR