Titriplex® III (ethylenedinitrilotetraacetic acid disodium salt dihydrate), EMPROVE® EXPERT ACS, JP, USP, BP, Ph. Eur. for biopharmaceutical production, SAFC®

Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate

1.37004.1000 1.37004.9012 1.37004.9025
1.37004.1000EA 656 EUR
1.37004.1000 1.37004.9012 1.37004.9025
Titriplex® III (ethylenedinitrilotetraacetic acid disodium salt dihydrate), EMPROVE® EXPERT ACS, JP, USP, BP, Ph. Eur. for biopharmaceutical production, SAFC®
EDTA disodium salt dihydrate
Formule: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
Smeltpunt: 255
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00150037
CAS nummer: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR