Magnesium chloride hexahydrate, EMPROVE® EXPERT JPC, USP, ACS, Ph. Eur., BP for biopharmaceutical production, SAFC®

Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Magnesium dichloride

1.37008.6025
1.37008.6025EA 0 EUR
1.37008.6025
Magnesium chloride hexahydrate, EMPROVE® EXPERT JPC, USP, ACS, Ph. Eur., BP for biopharmaceutical production, SAFC®
Magnesium chloride hexahydrate

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).


Certificaten: Corresponds to Ph Eur, BP, USP, JPC, ACS

Formule: MgCl₂·6H₂O
MW: 203.3 g/mol
Kookpunt: 1412
Smeltpunt: 117
Dichtheid: 1.57 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149781
CAS nummer: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684

Order Now

Specification Test Results

Assay (complexometric) 99.0 - 101.0 %
Identity passes test
Appearance of solution passes test
Insoluble matter ≤ 0.005 %
Acidity or alkalinity passes test
pH-value (5 %; water) 5.0 - 7.0
Bromide (Br) ≤ 0.05 %
Nitrate (NO3) (**) ≤ 0.001 %
Phosphate (PO4) (**) ≤ 0.0005 %
Sulfate (SO₄) ≤ 0.002 %
Heavy metals (as Pb) ≤ 0.0005 %
Al (Aluminium) ≤ 0.0001 %
As (Arsenic) ≤ 0.0002 %
Ba (Barium) passes test
Ba (Barium) (**) ≤ 0.005 %
Ca (Calcium) ≤ 0.01 %
Fe (Iron) ≤ 0.0005 %
Hg (Mercury) ≤ 0.0001 %
K (Potassium) ≤ 0.0050 %
Mn (Manganese) (**) ≤ 0.0005 %
Na (Sodium) (**) ≤ 0.005 %
NH₄ (Ammonium) ≤ 0.002 %
Pb (Lead) ≤ 0.0004 %
Sr (Strontium) (**) ≤ 0.005 %
Residual solvents (Ph Eur/USP/ICH) excluded by manufacturing process
Water 51.0 - 55.0 %
Endotoxins ≤ 3.0 I.U./g
Microbiological test (Total aerobic microbial count (TAMC)) ≤ 100 CFU/g
Microbiological test (Total yeast and mould count (TYMC)) ≤ 100 CFU/g
** additional requirements by ACS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR