D(-)-Mannitol, EMPROVE® EXPERT Ph. Eur., BP, USP, JP

Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

1.37096.1000 1.37096.9013 1.37096.9025 1.37096.9051
1.37096.1000EA 293 EUR
1.37096.1000 1.37096.9013 1.37096.9025 1.37096.9051
D(-)-Mannitol, EMPROVE® EXPERT Ph. Eur., BP, USP, JP
D(-)-Mannitol
Formule: HOH₂C(CH(OH))₄CH₂OH
Kookpunt: 290…295 °C (4 hPa)
Smeltpunt: 164…169 °C
Dichtheid: 1,49 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064287
CAS nummer: 69-65-8
EINECS: 200-711-8
Merck Index: 13,05769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163f556dca45086e05ff490ddc8eeb65d04aea05&vid=408e3f5566e309064ce68988cdd160305c5b8c0a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR