Polyvinyl alcohol, M.W. 115,000 ≥88%, EMPROVE® EXPERT JPE, Ph. Eur., USP suitable for use as excipient, hydrolysed, SAFC®

Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: PVOH, PVA, Mowiol®

1.41353.1000 1.41353.9029
1.41353.1000EA 542 EUR
1.41353.1000 1.41353.9029
Polyvinyl alcohol, M.W. 115,000 ≥88%, EMPROVE® EXPERT JPE, Ph. Eur., USP suitable for use as excipient, hydrolysed, SAFC®
Polyvinyl alcohol
Formule: [-CH₂CH(OH)-]n
MDL Nummer: MFCD00081922
CAS nummer: 9002-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR