Calcium chloride dihydrate, EMPROVE® ESSENTIAL E509, FCC, BP, Ph. Eur., JP, USP, SAFC®

Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Calcium Chloride Dihydrate, Calcium dichloride

1.42002.5000 1.42002.9029 1.42002.9525 1.42002.1000
1.42002.5000EA 288 EUR
1.42002.5000 1.42002.9029 1.42002.9525 1.42002.1000
Calcium chloride dihydrate, EMPROVE® ESSENTIAL E509, FCC, BP, Ph. Eur., JP, USP, SAFC®
Calcium chloride dihydrate

pH value: 4.5 - 8.5 (50 g/l, H₂O, 20 °C)
Solubility: 1280 g/l

Formule: CaCl₂·2H₂O
MW: 147.01 g/mol
Kookpunt: 1600
Smeltpunt: 176
Dichtheid: 1.85 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149613
CAS nummer: 10035-04-8
EINECS: 233-140-8
Merck Index: 13,01660

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR