HPLC columns, Hibar®, LiChrospher® 100 RP-8
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.00423.7087 1.00423.7088 1.00424.7088 1.50009.7088 1.50012.7087 1.50012.7088 1.50013.7088 1.50182.7088 1.50183.7087 1.50183.7088
1.00423.7087EA 0 EUR
1.00423.7087 1.00423.7088 1.00424.7088 1.50009.7088 1.50012.7087 1.50012.7088 1.50013.7088 1.50182.7088 1.50183.7087 1.50183.7088
HPLC columns, Hibar®, LiChrospher® 100 RP-8
Kolommen voor chromatografie
Stainless steel Hibar® columns are complete with endfittings. These columns are available with various types of LiChrospher® sorbents.
When using a guard column with a Hibar® RT column, we recommend part number 1.51487.0001 guard column cartridge holder for 4×4 mm guard column cartridges LiChroCART®.