HPLC columns, Hibar®, with Superspher® Sorbents
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.00423.7137 1.00423.7138 1.00423.7139 1.00423.7140 1.00423.7141 1.00423.7142 1.00424.7137 1.00424.7138 1.00424.7139 1.00424.7140 1.00424.7142 1.50009.7137 1.50009.7138 1.50009.7139 1.50009.7140 1.50009.7141 1.50009.7142 1.50012.7137 1.50012.7138 1.50012.7139 1.50012.7140 1.50012.7141 1.50012.7142 1.50013.7137 1.50013.7138 1.50013.7139 1.50013.7140 1.50013.7141 1.50013.7142 1.50182.7138 1.50182.7140 1.50182.7142 1.50183.7137 1.50183.7138 1.50183.7139 1.50183.7140 1.50183.7141 1.50183.7142 1.00423.7143
1.00423.7137EA 0 EUR
1.00423.7137 1.00423.7138 1.00423.7139 1.00423.7140 1.00423.7141 1.00423.7142 1.00424.7137 1.00424.7138 1.00424.7139 1.00424.7140 1.00424.7142 1.50009.7137 1.50009.7138 1.50009.7139 1.50009.7140 1.50009.7141 1.50009.7142 1.50012.7137 1.50012.7138 1.50012.7139 1.50012.7140 1.50012.7141 1.50012.7142 1.50013.7137 1.50013.7138 1.50013.7139 1.50013.7140 1.50013.7141 1.50013.7142 1.50182.7138 1.50182.7140 1.50182.7142 1.50183.7137 1.50183.7138 1.50183.7139 1.50183.7140 1.50183.7141 1.50183.7142 1.00423.7143
HPLC columns, Hibar®, with Superspher® Sorbents
Kolommen voor chromatografie
Stainless steel Hibar® columns are complete with endfittings. These columns are available with various types of Superspher® sorbents.
When using a guard column with a Hibar® RT column, we recommend part number 1.51487.0001 guard column cartridge holder for 4×4 mm guard column cartridges LiChroCART®.
Order Now
Specificaties
Environmentally Preferable