HPLC columns, LiChroCART®, Purospher® STAR NH₂
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.50229.7177 1.50195.7177 1.50234.7177 1.50201.7177 1.50177.7177 1.50179.7177 1.51431.7177 1.50174.7177 1.50175.7177 1.50228.7177 1.50233.7177 1.50236.7177 1.50302.7177 1.51442.7177 1.51443.7177 1.50170.7177 1.51432.7177 1.50173.7177 1.51444.7177 1.51445.7177 1.50171.7177 1.51448.7177 1.50178.7177
1.50229.7177EA 0 EUR
1.50229.7177 1.50195.7177 1.50234.7177 1.50201.7177 1.50177.7177 1.50179.7177 1.51431.7177 1.50174.7177 1.50175.7177 1.50228.7177 1.50233.7177 1.50236.7177 1.50302.7177 1.51442.7177 1.51443.7177 1.50170.7177 1.51432.7177 1.50173.7177 1.51444.7177 1.51445.7177 1.50171.7177 1.51448.7177 1.50178.7177
HPLC columns, LiChroCART®, Purospher® STAR NH₂
Kolommen voor chromatografie
Stainless steel LiChroCART® cartridges are available with various types of Purospher® sorbents.
These LiChroCART® cartridges require part number 1.51486.0001 manu-CART® cartridge column holder, which can be used to hold one cartridge column with or without a  4×4 mm guard column. LiChroCART® columns 250×10 mm require part number 1.51419.0001 manu-CART® 10. The short LiChroCART® columns can be ordered as a set including the corresponding cartridge holder and one cartridge, or as a pack of three cartridges without cartridge holder. The separate part numbers for the cartridge are asMore Product Information