Arsenic standard solution, 1000 mg/L As in dil. nitric acid (from H₃AsO₄), Certipur® standard for ICP, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1.70303.0100
1.70303.0100EA 183 EUR
1.70303.0100
Arsenic standard solution, 1000 mg/L As in dil. nitric acid (from H₃AsO₄), Certipur® standard for ICP, Supelco®
Standaarden Kationische standaarden Arseen standaarden
Certificaten: Traceable to SRM from NIST