Bovine serum albumin (BSA)

Leverancier: SEQENS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

1005-70-100G 1005-70-500G 1005-70-1KG
1005-70-100GEA 209 EUR
1005-70-100G 1005-70-500G 1005-70-1KG
Bovine serum albumin (BSA)
Eiwitten en peptiden

SEQENS IVD manufactures Bovine Serum Albumin (BSA) derived from plasma, collected from healthy animals of French origin. This product is obtained by 'Heat Shock' method, which ensures higher purity and preserves BSA intrinsic properties.


  • Albumin purity: >97%
  • Total proteins: >92%
  • Proteases: Undetectable
  • Shelf life: 3 years
  • Sterile filtered 0,2 µm prior to being freeze-dried


Full traceability and batch-to-batch repeatability guaranteed.


Leveringsinformatie: Supplied lyophilised.

Order Now

Specificaties

Protein/peptide name Bovine Serum Albumin
Environmentally Preferable
Protein synonyms BSA|Fraction V|Cohn Fraction V
Species Bovine
Bron Serum
Protease-free Y
Formulering lyophilized

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR