Antimony ≥99.8% (metals basis), rods, Length: 10 cm (3.9 in), Ø 12.7 mm (0.50 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

10098.KG 10098.KJ
10098.KGEA 81.8 EUR
10098.KG 10098.KJ
Antimony ≥99.8% (metals basis), rods, Length: 10 cm (3.9 in), Ø 12.7 mm (0.50 in)
Antimony

Purity: 99.8% CAS No.: 7440-36-0 Physical Form: ≈81.2g (0.178lbs)/10cm piece, Cast rod Odour: Odourless MDL No.: MFCD00134030

Formule: Sb
MW: 121.76 g/mol
Kookpunt: 1637
Smeltpunt: 630
Dichtheid: 6.684 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00134030
CAS nummer: 7440-36-0
EINECS: 231-146-5
UN: 2871
ADR: 6.1 III
Merck Index: 12,00733

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR