Antimony ≥99.5% (metals basis), powder -100 mesh

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

10100.30 10100.A1
10100.30EA 60.4 EUR
10100.30 10100.A1
Antimony ≥99.5% (metals basis), powder -100 mesh
Antimony

Purity: 99.5% CAS No.: 7440-36-0 Physical Form: Apparent density 2.4-2.9g/cm3 Odour: Odourless Boiling Point: 1380° UN No.: 2871 MDL No.: MFCD00134030

Formule: Sb
MW: 121.76 g/mol
Kookpunt: 1380 °C
Smeltpunt: 630
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00134030
CAS nummer: 7440-36-0
EINECS: 231-146-5
UN: 2871
ADR: 6.1 III
Merck Index: 12,00733

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR