Cadmium ≥99.95% (metals basis), tear drop shot -5/+20 mesh

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

10122.18 10122.30 10122.A1
10122.18EA 47.9 EUR
10122.18 10122.30 10122.A1
Cadmium ≥99.95% (metals basis), tear drop shot -5/+20 mesh
Cadmium

Purity: 99.95% CAS No.: 7440-43-9 MDL No.: MFCD00010914

Formule: Cd
Kookpunt: 765 °C (1013 hPa)
Smeltpunt: 321 °C
Dichtheid: 8,642 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010914
CAS nummer: 7440-43-9
EINECS: 231-152-8
UN: 2570
ADR: 6.1,II
Merck Index: 12,01649

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR