Zilvernitraat 99.8-100.5%, meets analytical specification of BP, Ph. Eur., USP, Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

10220-1KG 10220-500G 10220-100G 10220-50G
10220-1KGEA 3720 EUR
10220-1KG 10220-500G 10220-100G 10220-50G
Zilvernitraat 99.8-100.5%, meets analytical specification of BP, Ph. Eur., USP, Fluka™
Zilvernitraat
Formule: AgNO₃
Kookpunt: 444 °C (1013 hPa)
Smeltpunt: 212 °C
Dichtheid: 4,350 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003414
CAS nummer: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1,II
Bereik: 01-2119513705-43
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.8 - 100.5 %
Acid. or alk. react. impurities complying
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 0.0005 %
Lead (Pb) Max. 0.002 %
Substances not precipitated by HCl (as SO₄) Max. 0.03 %
Nitrite (NO₂) Max. 0.1 %
Sulfate (SO₄) Max. 0.01 %
Al, Pb, Bi and Cu ions complying
Appearance of the solution complying
Residual solvents complying
Vapor Density 5.8 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR