Platinum, wire for ISA type R or S standard grade thermocouple, Ø 0.0762 mm (0.003 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Platinium

10291.BW 10291.G2 10291.G6
10291.BWEA 37.5 EUR
10291.BW 10291.G2 10291.G6
Platinum, wire for ISA type R or S standard grade thermocouple, Ø 0.0762 mm (0.003 in)
Platinum

For ISA Type R or S Standard Grade Thermocouple


CAS No.: 7440-06-4 Physical Form: ≈0.05g/50cm Odour: Odourless MDL No.: MFCD00011179

Formule: Pt
MW: 195.08 g/mol
Kookpunt: 3827
Smeltpunt: 1769
Dichtheid: 21.447 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011179
CAS nummer: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR