Kalium ≥99.95% (metals basis), solid

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


Ampuled under argon

10299.06 10299.03
10299.06EA 364 EUR
10299.06 10299.03
Kalium ≥99.95% (metals basis), solid
Kalium

Purity: 99.95% CAS No.: 7440-09-7 Physical Form: Sealed in standard ampoules under vacuum Odour: Odourless UN No.: 2257 MDL No.: MFCD00133776

Formule: K
Kookpunt: 760 °C (1013 hPa)
Smeltpunt: 63 °C
Dichtheid: 0,86 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00133776
CAS nummer: 7440-09-7
EINECS: 231-119-8
UN: 2257
ADR: 4.3,I
Merck Index: 12,07763

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR