Caspase-3 substrates, NucView®

Leverancier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

10403 10402 10405-T 10405 10406 10406-T 10402-T 10403-T 10407 10407-T 10408 10408-T
10403.EA 583 EUR
10403. 10402. 10405-T 10405. 10406. 10406-T 10402-T 10403-T 10407. 10407-T 10408. 10408-T
Caspase-3 substrates, NucView®
Reagentia voor elektroforese Western Blotting Reagents

NucView® caspase-3 substrates are fluorescent probes that allow for the real-time detection of apoptosis by monitoring caspase-3/7 activity in intact cells using confocal microscopy, flow cytometry or microplate readers.


  • Bi-functional substrates
  • Rapid, no-wash assay for microscopy or flow cytometry
  • Stable for several days for performing time-course assays
  • Tolerant of formaldehyde fixation and permeabilisation
  • Use in adherent or suspension cell cultures
  • For flow cytometry, microscopy, live cell imaging, or microplate readers


NucView® substrates consist of a fluorogenic DNA dye coupled to the caspase-3/7 DEVD recognition sequence. Upon entering apoptotic cells, the non-fluorescent substrate is cleaved by caspases-3/7 releasing a high-affinity DNA binding dye which migrates to the nucleus and becomes fluorescent upon binding to DNA.


NucView® 488 caspase-3 substrate is offered as a 1 mM solution in PBS (phosphate-buffered saline) for cells that are sensitive to DMSO toxicity. NucView® 405 and 530 are available in 1 mM in PBS and trial sizes are available for both PBS and DMSO formulations.


Leveringsinformatie: Available in three colour options: NucView® 488: Ex/Em (bound to DNA) 500 nm/530 nm, NucView® 405 (bound to DNA): Ex/Em 429 nm/469 nm, NucView® 530 (bound to DNA): Ex/Em 528 nm/563 nm.


Opgelet: In non-DMSO sensitive cell types, adding DMSO during the substrate incubation may enhance NucView staining.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR