Tungsten ≥99.95% (metals basis), foil, Thickness: 0.5 mm (0.02 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

10414.FG 10414.FY 10414.GD 10414.GW 10414.HB
10414.FGEA 105 EUR
10414.FG 10414.FY 10414.GD 10414.GW 10414.HB
Tungsten ≥99.95% (metals basis), foil, Thickness: 0.5 mm (0.02 in)
Tungsten

Purity: 99.95% CAS No.: 7440-33-7 Physical Form: ≈12g/25x50mm MDL No.: MFCD00011461

Formule: W
Kookpunt: 5660 °C (1013 hPa)
Smeltpunt: 3410 °C
Dichtheid: 19 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011461
CAS nummer: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR