Tungsten ≥99.95% (metals basis), foil, Thickness: 0.25 mm (0.01 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

10415.FI 10415.GH 10415.HP
10415.FIEA 98.2 EUR
10415.FI 10415.GH 10415.HP
Tungsten ≥99.95% (metals basis), foil, Thickness: 0.25 mm (0.01 in)
Tungsten

Purity: 99.95% CAS No.: 7440-33-7 Physical Form: ≈12g/50x50mm MDL No.: MFCD00011461

Formule: W
Kookpunt: 5660 °C (1013 hPa)
Smeltpunt: 3410 °C
Dichtheid: 19 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011461
CAS nummer: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR