Zink ≥99.9% (metals basis), foil, Width: 30 cm (12 in), Thickness: 0.62 mm (0.024 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

10437.CB 10437.BW
10437.CBEA 308 EUR
10437.CB 10437.BW
Zink ≥99.9% (metals basis), foil, Width: 30 cm (12 in), Thickness: 0.62 mm (0.024 in)
Zink

Purity: 99.9% CAS No.: 7440-66-6 Physical Form: ≈665g/30x50cm Odour: Odourless MDL No.: MFCD00011291

Formule: Zn
Kookpunt: 908 °C (1013 hPa)
Smeltpunt: 420 °C
Dichtheid: 7,14 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011291
CAS nummer: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR