Palladium(II) acetate (45,9 - 48,4% Pd)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Palladium diacetate, Palladium (II) acetate

10516.03 10516.06 10516.14
10516.03EA 164 EUR
10516.03 10516.06 10516.14
Palladium(II) acetate (45,9 - 48,4% Pd)
Palladium(II) acetate

CAS No.: 3375-31-3 Molecular Formula: C12H18O12Pd3 Formula Weight: 673.46 Physical Form: Needles Appearance: Red-brown Melting Point: ca 205° dec. MDL No.: MFCD00012453


Carbonylation, oxidation and C-C bond formation

Formule: C₁₂H₁₈O₁₂Pd₃
MW: 673.46 g/mol
Smeltpunt: ∼ 205 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012453
CAS nummer: 3375-31-3
EINECS: 222-164-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR