Indium ≥99.9995% (metals basis), shot, Puratronic® ≤ 5 mm (0.2 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

10613.09 10613.18
10613.09EA 132 EUR
10613.09 10613.18
Indium ≥99.9995% (metals basis), shot, Puratronic® ≤ 5 mm (0.2 in)
Indium

Purity: 99.9995% CAS No.: 7440-74-6 Odour: Odourless MDL No.: MFCD00134048

Formule: In
MW: 114.82 g/mol
Kookpunt: 2000
Smeltpunt: 156
Dichtheid: 7.3 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00134048
CAS nummer: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR