Aluminium oxide ≥99.995% (metals basis), Puratronic®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

10627.14 10627.22 10627.36
10627.14EA 169 EUR
10627.14 10627.22 10627.36
Aluminium oxide ≥99.995% (metals basis), Puratronic®
Aluminium oxide

Purity: 99.995% CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: Powder Density: 3.965 Refractive Index: 1.768 Odour: Odourless Melting Point: 2045° Boiling Point: 2980° Merck Reference: 14,356 MDL No.: MFCD00003424

Formule: Al₂O₃
Kookpunt: 2980 °C
Smeltpunt: 2045 °C
Dichtheid: 3,965
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003424
CAS nummer: 1344-28-1
EINECS: 215-691-6
Bereik: 01-2119529248-35
Merck Index: 14,356

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR