Barium chloride dihydrate ≥99.997% (metals basis), Puratronic®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Barium dichloride

10646.14 10646.22 10646.36
10646.14EA 74.2 EUR
10646.14 10646.22 10646.36
Barium chloride dihydrate ≥99.997% (metals basis), Puratronic®
Barium chloride dihydrate

Purity: 99.997% CAS No.: 10326-27-9 Molecular Formula: BaCl2.2H2O Formula Weight: 244.28 (208.25anhy) Physical Form: Powder Density: 3.097 Appearance: White Odour: Odourless Melting Point: 113° -2H2O Merck Reference: 14,971 UN No.: 1564 MDL No.: MFCD00149154

Formule: BaCl₂·2H₂O
MW: 244.27 g/mol
Kookpunt: 1560
Smeltpunt: 113 °C (-2H₂O)
Dichtheid: 3.097
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149154
CAS nummer: 10326-27-9
EINECS: 600-412-6
UN: 1564
ADR: 6.1 III
Merck Index: 14,971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR