Bismuth(III) iodide ≥99.999% (metals basis), Puratronic®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Bismuth triiodide

10656.04 10656.09 10656.18
10656.04EA 45.9 EUR
10656.04 10656.09 10656.18
Bismuth(III) iodide ≥99.999% (metals basis), Puratronic®
Bismuth(III) iodide

Purity: 99.999% CAS No.: 7787-64-6 Molecular Formula: BiI3 Formula Weight: 589.69 Physical Form: Lump/Powder Density: 5.778 Appearance: Dark grey to black Odour: Acidic Melting Point: 408° Boiling Point: ca 500° Merck Reference: 14,1269 UN No.: 3260 MDL No.: MFCD00010894

Formule: BiI₃
Kookpunt: ∼500 °C
Smeltpunt: 408 °C
Dichtheid: 5,778
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010894
CAS nummer: 7787-64-6
EINECS: 232-127-4
UN: 3260
ADR: 8,III
Merck Index: 14,1269

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR