Chromium(III) oxide ≥99.97% (metals basis), powder, Puratronic® -22 Mesh

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

10688.06 10688.14 10688.22 10688.36
10688.06EA 61.7 EUR
10688.06 10688.14 10688.22 10688.36
Chromium(III) oxide ≥99.97% (metals basis), powder, Puratronic® -22 Mesh
Chromium(III) oxide

Purity: 99.97% CAS No.: 1308-38-9 Molecular Formula: Cr2O3 Formula Weight: 151.99 Physical Form: -22 Mesh Powder Density: 5.22 Refractive Index: 2.551 Appearance: Pale to dark green Odour: Odourless Melting Point: 2435° Boiling Point: 3000° Merck Reference: 14,2234 MDL No.: MFCD00010949

Formule: Cr₂O₃
Kookpunt: 3000 °C
Smeltpunt: 2435 °C
Dichtheid: 5,22
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010949
CAS nummer: 1308-38-9
EINECS: 215-160-9
Merck Index: 14,2234

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR