Cobalt(II,III) oxide ≥99.9985% (metals basis), Puratronic®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Tricobalt tetraoxide, Cobaltic tetraoxide

10695.09 10695.18
10695.09EA 130 EUR
10695.09 10695.18
Cobalt(II,III) oxide ≥99.9985% (metals basis), Puratronic®
Cobalt(II,III) oxide

Purity: 99.9985% CAS No.: 1308-06-1 Molecular Formula: Co3O4 Formula Weight: 240.80 Physical Form: Powder Density: 6.11 Appearance: Black Melting Point: 900° dec. Merck Reference: 14,2429 UN No.: 3077 MDL No.: MFCD00010939

Formule: Co₃O₄
MW: 240.8 g/mol
Smeltpunt: 900 °C (ontbinding)
Dichtheid: 6.11
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010939
CAS nummer: 1308-06-1
EINECS: 215-157-2
UN: 3077
ADR: 9 III
Merck Index: 14,2429

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR