Strontium fluoride ≥99.99% (metals basis), Puratronic®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

10878.06 10878.14 10878.22
10878.06EA 65.7 EUR
10878.06 10878.14 10878.22
Strontium fluoride ≥99.99% (metals basis), Puratronic®
Strontium fluoride

Purity: 99.99% CAS No.: 7783-48-4 Molecular Formula: SrF2 Formula Weight: 125.62 Physical Form: Powder Density: 4.240 Refractive Index: 1.442 Appearance: White Odour: Odourless Melting Point: 1473° Merck Reference: 14,8842 MDL No.: MFCD00011251

Formule: F₂Sr
Kookpunt: 2460 °C (1013 hPa)
Smeltpunt: 1473 °C
Dichtheid: 4,240
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011251
CAS nummer: 7783-48-4
EINECS: 232-000-3
UN: 3288
ADR: 6.1,III
Merck Index: 14,8842

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR