Platinum ≥99.997% (metals basis), draad, Premion®, Ø 0.25 mm (0.01 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

10958.BS 10958.BW 10958.CB
10958.BSEA 98 EUR
10958.BS 10958.BW 10958.CB
Platinum ≥99.997% (metals basis), draad, Premion®, Ø 0.25 mm (0.01 in)
Platinum

Purity: 99.997% CAS No.: 7440-06-4 Physical Form: ≈1.05g/m Odour: Odourless MDL No.: MFCD00011179

Formule: Pt
Kookpunt: 3827 °C (1013 hPa)
Smeltpunt: 1769 °C
Dichtheid: 21,447 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011179
CAS nummer: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR