Palladium ≥99.99% (metals basis), draad, Premion®, Ø 0.25 mm

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

10962.G1 10962.G5 10962.G9
10962.G1EA 199 EUR
10962.G1 10962.G5 10962.G9
Palladium ≥99.99% (metals basis), draad, Premion®, Ø 0.25 mm
Palladium

Purity: 99.99% CAS No.: 7440-05-3 Physical Form: ≈0.6g/m Odour: Odourless MDL No.: MFCD00011167

Formule: Pd
Kookpunt: 3167 °C (1013 hPa)
Smeltpunt: 1555 °C
Dichtheid: 12,02 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011167
CAS nummer: 7440-05-3
EINECS: 231-115-6
Merck Index: 12,07121

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR