Copper ≥99.999% (metals basis), wire, Puratronic®, Ø 0.5 mm (0.02 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

10973.G5 10973.G9
10973.G5EA 148 EUR
10973.G5 10973.G9
Copper ≥99.999% (metals basis), wire, Puratronic®, Ø 0.5 mm (0.02 in)
Copper

Purity: 99.999% CAS No.: 7440-50-8 Physical Form: ≈1.75g/m Odour: Odourless MDL No.: MFCD00010965

Formule: Cu
Kookpunt: 2595 °C (1013 hPa)
Smeltpunt: 1083 °C
Dichtheid: 8,94 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010965
CAS nummer: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR