Tellurium ≥99.99% (metals basis), powder -30 mesh

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

11074.14 11074.22
11074.14EA 104 EUR
11074.14 11074.22
Tellurium ≥99.99% (metals basis), powder -30 mesh
Tellurium

Purity: 99.99% CAS No.: 13494-80-9 Odour: Odourless UN No.: 3288 MDL No.: MFCD00134062

Formule: Te
Kookpunt: 990 °C (1013 hPa)
Smeltpunt: 450 °C
Dichtheid: 8,83 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00134062
CAS nummer: 13494-80-9
EINECS: 236-813-4
UN: 3288
ADR: 6.1,III
Merck Index: 12,09273

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR