Tellurium ≥99.9999% (metals basis), pieces, zone refined

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

11076.18 11076.30 11076.09
11076.18EA 284 EUR
11076.18 11076.30 11076.09
Tellurium ≥99.9999% (metals basis), pieces, zone refined
Tellurium

Purity: 99.9999% CAS No.: 13494-80-9 Odour: Odourless MDL No.: MFCD00134062

Formule: Te
Kookpunt: 990 °C (1013 hPa)
Smeltpunt: 450 °C
Dichtheid: 8,83 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00134062
CAS nummer: 13494-80-9
EINECS: 236-813-4
UN: 3284
ADR: 6.1,III
Merck Index: 12,09273

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR