Safety helmets, uvex pheos alpine
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9773.350 9773.050 9773.150 9773.950 9773.550 9773.250 9773.450
111-2277EA 139 EUR
111-2277 111-2276 111-1917 111-1928 111-1927 111-1918 111-1919
Safety helmets, uvex pheos alpine
Hoofdbescherming Veiligheidshelmen
This light ABS helmet offers a high level of protection and durability. It is multifunctional and combines features from industrial protection helmets (EN 397) and requirements for climbing helmets (EN 12492).
  • Side Euroslot adapters (30 mm) for attaching earmuffs
  • Attachment for face protection available
  • Six-point textile suspension harness ensures optimum fit and comfort
  • Clips for attaching extras with elastic band, e.g. wide vision goggles and LED climbing helmet torch
  • Head torches (miner’s lamp) can be attached
  • Suitable for ambient temperatures down to −3
More Product Information