Zirconium(IV) oxide ≥99.978% (metals basis), Puratronic®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Zirconium dioxide, Zirconium Oxide

11141.14 11141.22 11141.36
11141.14EA 238 EUR
11141.14 11141.22 11141.36
Zirconium(IV) oxide ≥99.978% (metals basis), Puratronic®
Zirconium(IV) oxide

Purity: 99.978% CAS No.: 1314-23-4 Molecular Formula: O2Zr Formula Weight: 123.22 Physical Form: Powder Density: 5.89 Appearance: White Odour: Odourless Melting Point: ca 2700° Boiling Point: ca 5000° Merck Reference: 14,10180 MDL No.: MFCD00011310

Formule: ZrO₂
MW: 123.22 g/mol
Kookpunt: ∼5000 °C
Smeltpunt: ∼2700 °C
Dichtheid: 5.89
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011310
CAS nummer: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR