Germanium ≥99.9999% (metals basis), pieces, Puratronic® ≤ 2 cm (0.8 in)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

11144.06 11144.14 11144.22
11144.06EA 129 EUR
11144.06 11144.14 11144.22
Germanium ≥99.9999% (metals basis), pieces, Puratronic® ≤ 2 cm (0.8 in)
Germanium

Purity: 99.9999+% CAS No.: 7440-56-4 Physical Form: Electrical Resistivity: 40 ohm-cm Odour: Odourless MDL No.: MFCD00085310

Formule: Ge
Kookpunt: 2830 °C (1013 hPa)
Dichtheid: 5,323 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00085310
CAS nummer: 7440-56-4
EINECS: 231-164-3
Merck Index: 12,04415

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR