Snijbestendige kettinghandschoenen, Chainex 2000
Leverancier: HONEYWELL SAFETY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2500000R0302 2500001R0302 2500002R0302 2500003R0302 2500004R0302 2500005R0302
112-1243EA 135 EUR
112-1243 112-1244 112-1249 112-1255 112-1256 112-1257
Snijbestendige kettinghandschoenen, Chainex 2000
Handschoenen
The Chainex 2000 is a full stainless steel glove with short cuff and attached textile strap. Chainex 2000 is a fully reversible product range, and is equipped with heavy duty adjustable straps and press stud fastening. Offers excellent cut and puncture protection. The high quality of the chain mesh reduces the numbers of repairs, and extends the life time of the glove.
  • Smooth textile straps adapt to all wrist profiles
  • Fully reversible gloves can be used on left or right hand
  • Wrist fastens with press stud and adjustable buckle
Stainless chainmailMore Product Information