Nitril handschoenen, TouchNTuff® 92-600 / 92-605

Leverancier: Ansell
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

92-600/8.5-9 92-600/7.5-8 92-600/6.5-7 92-600/9.5-10 92-605/S 92-605/M 92-605/L 92-605/XL 92-600/10.5-11 92-600/5.5-6
112-0999EA 38.1 EUR
112-0999 112-0998 112-0997 112-1008 112-1935 112-1936 112-1937 112-1938 112-6640 112-6641
Nitril handschoenen, TouchNTuff® 92-600 / 92-605
Handschoenen
These gloves with rolled cuffs are made of puncture-resistant nitrile, which offers valuable protection against accidental splashes of hazardous chemicals.

  • Non sterile, powder-free, smooth finish or textured fingertips
  • 100% nitrile; no waxes, silicone, or plasticisers
  • Ideal for sensitive skin
  • Three times the puncture resistance of latex or PVC
  • Light, cool, and durable
  • Designed for maximum freedom of movement and minimum hand fatigue

Ideaal voor gebruik in de chemische en elektronische industrie.

Thickness: 0,12 mm

Colour: Green

EN 374, EN 1149More Product Information
New Product
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR