Cleanroom handschoenen, nitril, BioClean Nerva™
Leverancier: Nitritex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BNAL-L BNAL-M BNAL-S BNAL-XL
112-3848EA 1030 EUR
112-3848 112-3849 112-3850 112-3851
Cleanroom handschoenen, nitril, BioClean Nerva™
Handschoenen
These ambidextrous nitrile gloves with beaded cuffs provide good resistance against chemicals. They also have excellent ESD properties.
  • Non sterile, powder-free, textured fingertips
  • Contain no natural latex proteins, hypoallergenic
  • Extended length provides protection of the forearm
  • Extended leaching in pure deionised water ensures extremely low levels of chlorides and extractable metals
  • Textured fingers provide enhanced grip
Geschikt voor cleanrooms Klasse 10, ISO 4 of M2.5.
Certificaten: Manufactured in a facility holding ISO 9001:2015 and (EU)2016More Product Information