Chemical resistant gloves, Tychem® NP570 CT
Leverancier: DuPont
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
NP5700BK080144
112-5468EA 0 EUR
112-5468
Chemical resistant gloves, Tychem® NP570 CT
Handschoenen
These gloves are made of a yarn liner with neoprene coating and provide ANSI level A5 and EN level 5 cut protection, as well as first-class defence against acids, caustics, solvents, greases and oils.
  • 13-gauge seamless cut-resistant engineered knit liner for maximum comfort and endurance
  • Flexible coating
  • Tactile non-slip grip
Ideal protection where tasks present the dual risk of chemical exposure and cut.
Certificaten: EN 388:2016 (3X43E), EN ISO 374-1:2016 Type A (AJKLMT), EN ISO 374-5:2016 (VIRUS); ANSI cut level A5; Cat. III.