General purpose gloves, Showa 7166R
Leverancier: Showa
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
7166R-10
112-5892EA 13 EUR
112-5892
General purpose gloves, Showa 7166R
Handschoenen
The 7166 general purpose gloves feature an impermeable full nitrile coating that protects hands from a range of hazards, including abrasion, oils, and chemicals.
  • Reinforced safety cuff prevents liquid and debris from entering the glove
  • Soft cotton liner controls the temperature and wicks away sweat
  • Rough surface grip is ideal for wet or greasy environments and increased protection
Typical applications include: Chemical spray and treatment, cutting roses and thorns, public utility, washing and cleaning, drainage, piping, guttering and drain pipe,More Product Information
Order Now
Specificaties
Model 7166R
Ambidexter/handspecifiek Handspecifiek
Environmentally Preferable
Voor eenmalig gebruik/herbruikbaar Herbruikbaar
Liner material Katoenen jersey
Finish Full nitrile coating
Cuff style Veiligheidsmanchet
EN-norm EN 388:2016 (4111X)