Life science nitrile gloves, PUREZERO® MARIN-XTRA®
Leverancier: Halyard
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
LFS121LG LFS121MD LFS121SM LFS121XL LFS121XS
112-6273EA 17.9 EUR
112-6273 112-6272 112-6271 112-6274 112-6270
Life science nitrile gloves, PUREZERO® MARIN-XTRA®
Handschoenen
Extended length PUREZERO® life science nitrile gloves deliver on all fronts. They are tested against 57 chemicals, with 3 levels of colour-coded protection offering increasing thickness designed for life science applications. The SAFESKIN® manufacturing facility uses 92% renewable energy, dispensers are made with up to 85% recycled materials, and cases are made with up to 99% recycled pulp.
  • Barrier protection (57 chemicals, 14 chemotherapy drugs)
  • Good fit and comfort
  • Textured fingertips provide tactile agility
  • Beaded cuff allows for ease of donning
  • Not
More Product Information