Life science nitrile gloves, PUREZERO® ULTRA-VIOLET®
Leverancier: Halyard
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
LFS511LG LFS511MD LFS511SM LFS511XL LFS511XS
112-6288EA 32 EUR
112-6288 112-6287 112-6286 112-6289 112-6285
Life science nitrile gloves, PUREZERO® ULTRA-VIOLET®
Handschoenen
PUREZERO® life science nitrile gloves deliver on all fronts. They are tested against 57 chemicals, with 3 levels of colour-coded protection offering increasing thickness designed for life science applications. The dispensers are made with up to 85% recycled materials, and cases are made with up to 99% recycled pulp.
  • Barrier protection (57 chemicals, 14 chemotherapy drugs)
  • Good fit and comfort
  • Textured fingertips provide tactile agility
  • Beaded cuff allows for ease of donning
  • Not made with natural rubber latex, for protection against Type IV allergies
  • Supply
More Product Information