Scandium(III) chloride hexahydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

11219.03 11219.06
11219.03EA 201 EUR
11219.03 11219.06
Scandium(III) chloride hexahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Scandium(III) chloride hexahydrate

Purity: 99.99% CAS No.: 20662-14-0 Molecular Formula: ScCl3.6H2O Formula Weight: 259.41 (151.32anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Merck Reference: 14,8392 MDL No.: MFCD00149852

Formule: Cl₃Sc·6H₂O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149852
CAS nummer: 20662-14-0
EINECS: 233-799-1
Merck Index: 14,8392

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR