Lanthanum(III) oxide ≥99.99% (REO, rare earth oxide basis), REacton®

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

11264.30 11264.18 11264.A1
11264.30EA 170 EUR
11264.30 11264.18 11264.A1
Lanthanum(III) oxide ≥99.99% (REO, rare earth oxide basis), REacton®
Lanthanum(III) oxide

Purity: 99.99% CAS No.: 1312-81-8 Molecular Formula: La2O3 Formula Weight: 325.82 Physical Form: Powder Density: 6.51 Melting Point: 2307° Boiling Point: 4200° Merck Reference: 14,5363 MDL No.: MFCD00011071

Formule: La₂O₃
MW: 325.81 g/mol
Kookpunt: 4200 °C
Smeltpunt: 2307 °C
Dichtheid: 6.51
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011071
CAS nummer: 1312-81-8
EINECS: 215-200-5
Merck Index: 14,5363

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR