Chemisch bestendige overalls, Tychem® 6000 F, modellen CHA5/CHA6
Leverancier: DuPont
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TFCHA5TGYXL002 TFCHA5TGY2X002 CHA5TGY16/XL CHA5TGY16/XXL CHA5TGY16/XXXL CHA5TOR00/XL CHA5TOR00/XXL TFCHA5TGYLG002 TFCHA5TGYMD002 TFCHA5TGYSM002 TYFCHA5TGY16/L TYFCHA5TGY16/M TYFCHA5TGY16/S TYFCHA5TOR00/L TYFCHA5TOR00/M TFCHA5TGY3X002 CHA5TOR00/3XL CHA5TOR00/4XL CHA5TOR00/5XL
113-0531EA 1380 EUR
113-0531 113-0532 113-0533 113-0534 113-0535 113-1744 113-1771 113-0536 113-0537 113-0538 113-0539 113-0540 113-0541 113-1897 113-1937 113-1468 DUPOCHA5TOR00/3XL DUPOCHA5TOR00/4XL DUPOCHA5TOR00/5XL
Chemisch bestendige overalls, Tychem® 6000 F, modellen CHA5/CHA6
Wegwerpkleding
Tychem® 6000 F overalls are made from a lightweight and durable fabric (<500 g per garment) consisting of a proprietary barrier film laminated to a heavy duty Tyvek® substrate. They offer excellent chemical permeation protection to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
  • With hood; elasticated face, wrists, waist and ankles for an optimal fit
  • Self-adhesive
More Product Information