Chemical resistant overalls, Tychem® 4000 S, model CHZ5 / CHZ6
Leverancier: DuPont
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SLCHZ5TWH00/L SLCHZ5TWH00/M SLCHZ5TWH00/S SLCHZ5TWH00/XL SLCHZ5TWH003XL SLCHZ5TWH00XXL SLCHZ6TWH16/L SLCHZ6TWH16/M SLCHZ6TWH16/S SLCHZ6TWH16/XL SLCHZ6TWH163XL SLCHZ6TWH16XXL
113-7968EA 983 EUR
113-7968 113-7967 113-7966 113-7969 113-7971 113-7970 113-7974 113-7973 113-7972 113-7975 113-7977 113-7976
Chemical resistant overalls, Tychem® 4000 S, model CHZ5 / CHZ6
Wegwerpkleding
These garments are made from a lightweight, supple and durable fabric (<700 g per garment) consisting of a chemical barrier film laminated to a heavy duty Tyvek® substrate. They achieve a barrier to permeation by a broad range of inorganic and organic chemicals.
  • Hood, double zips and double flaps for greater tightness, outer flap with adhesive tape and integrated chin flap
  • Elasticated face, wrists, waist and ankles for an optimal fit, thumb loops to prevent sleeves from riding up
  • Stitched and over-taped seams with barrier tape for protection and
More Product Information