Quantus® Protect Xtra, Cleanroom Coats
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
113-9942EA 0 EUR
113-9942 113-9943 113-9944 113-9945 113-9946 113-9947 113-9948 113-9949 113-9950
Quantus® Protect Xtra, Cleanroom Coats
Werkkleding
These coats are made from Protect Xtra fabric which is a further development of the Protect fabric consisting of 100% polyester weave with conductive yarn (warp). This fabric has been especially designed for use in critical areas (e.g. operating rooms, sterile environments). Thanks to the PTFE finish, the coats provide very good splash protection against liquids and other contamination. The fabric has good tensile strength and provides very good resistance to microbial penetration.
  • Lapped seams ensure well-sealed and secure seams
  • Press-stud closure, high
More Product Information